Cole-Parmer Reusable Low-Temperature Labels, Freezer, -5 to 30F (-21 to -1C); 10/Pk
Representative image only.
Cole-Parmer Reusable Low-Temperature Labels, Freezer, -5 to 30F (-21 to -1C); 10/Pk
Cole-Parmer | Item: 9030885
Ideal for use in refrigerator, freezer and cooler applications
₹5,300.00INR / Pkg of 10
₹5,300.00INR / Pkg of 10
Product Description
Key Features
  • Nontoxic nonhazardous temperature-measuring alternative
  • 1 year warranty
More About this Item
These self-adhesive reusable temperature labels provide a safe, accurate, easy-to-read means for monitoring low temperatures. Indicating strips use liquid crystal technology to display the current temperature range at an economical cost. Shelf life: 1 year after date of purchase.
1 Year Warranty
Specifications
  • Number of Temperature Indicators1
  • Max temperature (° F)30
  • Max temperature (° C)-1
  • Temperature Points (° F)(-5) to 30
  • Temperature Points (° C)(-21) to (-1)
  • Height (in)5 1/2
  • Width (in)1
  • DescriptionReusable Low-Temperature Labels, Freezer, -5 to 30F (-21 to -1C); 10/Pk