Cole-Parmer Replacement Silencer for Vacuum Pumps 78165-10 and -20
Representative image only.
Cole-Parmer Replacement Silencer for Vacuum Pumps 78165-10 and -20
Cole-Parmer | Item: 7816552
Reduce vacuum pump noise with minimal resistance to air flow
₹5,700.00INR / Each
₹5,700.00INR / Each
Product Description
Key Features
  • Increase productivity and operator safety—reduces irritating noise, improves working environment
  • Maintains clean environment
  • Reduces maintenance costs
  • Increases equipment life
  • Compact, lightweight, and durable
  • 1/4" NPT(M) threaded connection
  • Nylon body with felt element
  • 5 year warranty
More About this Item
Silencers, filters, regulators, and oils keep your vacuum pump at its best.
Please note: There will be a muffler change in the 2nd half of 2022. The vacuum pump may ship with a slightly different muffler and an increase noise level of 6 dB.
5 Year Warranty
Specifications
  • MaterialNylon body with felt element
  • Compatible WithVacuum pumps 78165-10 and -20
  • Connections1/4" NPT(M)
  • Length (InchShort)1 3/4
  • Diameter (InchShort)3/4
  • Length (cm)4.3
  • Diameter (cm)2
  • DescriptionReplacement Silencer for Vacuum Pumps 78165-10 and -20
  • Warranty5 years