Cole-Parmer Replacement Silencer for Vacuum Pump 78165-30
Representative image only.
Cole-Parmer Replacement Silencer for Vacuum Pump 78165-30
Cole-Parmer | Item: 7816554
Reduce vacuum pump noise with minimal resistance to air flow
₹6,900.00INR / Each
₹6,900.00INR / Each
Product Description
Key Features
  • Increase productivity and operator safety—reduces irritating noise, improves working environment
  • Maintains clean environment
  • Reduces maintenance costs
  • Increases equipment life
  • Reliable, straight through design—non-clogging
  • Acetal body with porous polyethylene element
  • 5 year warranty
More About this Item
Silencers, filters, regulators, and oils keep your vacuum pump at its best.
5 Year Warranty
Specifications
  • Compatible WithVacuum pump 78165-30
  • Connections1/4" NPS(M)
  • Length (InchShort)2 3/16
  • Diameter (InchShort)3/4
  • Length (cm)5.5
  • Diameter (cm)2
  • DescriptionReplacement Silencer for Vacuum Pump 78165-30
  • Warranty5 Years