Cole-Parmer Replacement Pestle/Mixer Adapter for Motor, 1 each
Representative image only.
Cole-Parmer Replacement Pestle/Mixer Adapter for Motor, 1 each
Cole-Parmer | Item: 4446826
Quickly homogenize cells, plant tissue, and DNA pellets within microtube vials
₹3,200.00INR / Each
₹3,200.00INR / Each
Buy It Together
Current Item
Cole-Parmer Replacement Pestle/Mixer Adapter for Motor, 1 each
Cole-Parmer Replacement Pestle/Mixer Adapter for Motor, 1 each
4446826
₹3,200.00INR / Each
+ +
Cole-Parmer Pestle and 1.5 mL Microtube, 100/pk
4446823
₹2,700.00INR / Pkg of 100
=
Total Price 12000.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Use with available microtubes free of DNAse, RNAse, and pyrogens to eliminate sample contamination
More About this Item
Use the battery-operated motor mixer with pestles and microtubes to drastically reduce hand fatigue when mixing multiple samples. Cell tissue and pellets are completely homogenized within seconds by means of high-speed vortexing action. Order pestles and tubes separately. The replacement rubber adapter, model 44468-26, is the adapter only, and is only compatible with the 44468-25 pestle mixer. This adapter atteches the motorized mixer to the pestle connector.
Specifications
  • MaterialSilicone
  • DescriptionReplacement Pestle/Mixer Adapter for Motor