Cole-Parmer Replacement ORP Electrode Cartridge for 27106-50
Representative image only.
Cole-Parmer Replacement ORP Electrode Cartridge for 27106-50
Cole-Parmer | Item: 2710662
Keep original housing and simply replace the electrode cartridge
₹66,200.00INR / Each
₹66,200.00INR / Each
Product Description
Key Features
  • Fluid flow across the surface provides a cleaning action that extends the life of the electrode and improves performance Flat measuring surface minimizes electrode coating, scratching, and breakage
  • 1 year warranty
More About this Item
Perfect for systems with turbulent flow. Includes electrode mounting glands with 3⁄4" NPT(M) threads.
1 Year Warranty
Specifications
  • Electrode TypeDouble junction
  • Electrode Body MaterialCPVC
  • Max Temperature (° C)100
  • DescriptionReplacement ORP Electrode Cartridge for 27106-50