Cole-Parmer Ratchet-Style Pinch Clamp, Polypropylene, Large; 25/Pk
Representative image only.
Cole-Parmer Ratchet-Style Pinch Clamp, Polypropylene, Large; 25/Pk
Cole-Parmer | Item: 5013113
Shut-off the flow with a simple push
₹5,300.00INR / Pkg of 25
₹5,300.00INR / Pkg of 25
Product Description
Key Features
  • Lightweight and reusable
  • Rust and corrosion resistant
  • USP Class VI and FDA 21 CFR 177.1520 compliant—excellent for biotechnology and pharmaceutical applications for transferring sterile fluids
  • Sterilize by autoclave or gamma irradiation
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
More About this Item
Ratchet-style pinch clamps provide an easy way to control the flow in flexible tubing. Multi-position ratchet control adjusts to allow flow from full flow to complete stop; simply push down all the way until it latches to stop the fluid flow.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Clamp MaterialPolypropylene
  • Clamp TypePinch
  • ColorWhite
  • Max Grip Range (InchShort)0.75
  • Width (in)1
  • Length (in)2.3125
  • DescriptionRatchet-Style Pinch Clamp, Polypropylene, Large; 25/Pk
  • WarrantyManufacturer defect