Cole-Parmer PVac™ Portable Vacuum System

Representative image only.

Cole-Parmer PVac™ Portable Vacuum System

Cole-Parmer | Item: 0439707

Construct your own waste aspiration system—no other vacuum source needed!

Call For Price

Call For Price

Buy It Together

Current Item

Cole-Parmer PVac™ Portable Vacuum System
Cole-Parmer PVac™ Portable Vacuum System
0439707

Call For Price
+ + + =
Total Price 223100.00 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Vacuum pump, HandE-Vac handheld aspirator with single-channel pipette tip adapter, inline filter, silicone vacuum tubing, rechargeable lithium battery, and universal power supply.

Key Features

  • Take into the field and run off the rechargeable battery to filter samples on site
  • Compact, portable liquid waste disposal device accommodates various collection vessels
  • Operate in the lab on the universal power adapter—power supply also acts as battery charger
  • Hydrophobic filter protects pump from over-filling
  • 3 year warranty

More About this Item

Combine this oil-free, diaphragm vacuum pump with a collection bottle to create a portable waste aspiration system and rapidly remove supernatant from a variety of cell culture or multiwell plates and dishes. The vacuum system can also be used as a stand-alone pump for vacuum-assisted filtration applications like bottletop and vacuum manifold filtration.

Compact size makes the vacuum system easily portable and allows for placement in an individual workstation or under a hood. A 0.45-µm hydrophobic, in-line filter protects from overfilling or fluid aspiration into the pump—ideal for filtering hazardous samples too dangerous for house lines, and as the vacuum source for waste aspiration systems. Couple with any glass or plastic collection bottle or flask*—like the Mini-Vac waste collection bottle—and empty a 2-liter bottle of waste in 40 seconds!

The HandE-Vac handheld aspirator (included) lets you control the vacuum pressure and speed by manually adjusting aperture button. An adapter for a single channel pipette is included; order different adapters for aspiration of multiwell plates and Pasteur pipettes.
*Collection bottle used with the P-Vac vacuum system should not exceed 2 liters.

3 Year Warranty
Specifications
  • Max Flow Rate (CFM)0.14
  • Max Flow Rate (Liters/min)4
  • Max Vacuum (in Hg)14
  • Max Vacuum (mbar)475
  • Power (VAC)100 to 240
  • Motor TypeDiaphragm
  • Stages1
  • Height (in)5.75
  • Depth (in)5.75
  • Width (in)5
  • Height (cm)14.7
  • Depth (cm)14.7
  • Width (cm)12.7
  • DescriptionPortable Vacuum System
  • Warranty3 Years
Reviews
Q&A