Cole-Parmer PTFE Magnet Retriever, Rare Earth, 450 x 10 mm
Representative image only.
Cole-Parmer PTFE Magnet Retriever, Rare Earth, 450 x 10 mm
Cole-Parmer | Item: 7890343
For the removal of stirrer magnets from vessels of all kinds
₹6,800.00INR / Each
₹6,800.00INR / Each
Product Description
Key Features
  • PTFE construction
More About this Item
Use with regular or rare earth stir bar models.
Specifications
  • DescriptionPTFE Magnet Retriever, Rare Earth, 450 x 10 mm