Cole-Parmer PTFE Beaker Cover for 06300-78
Representative image only.
Cole-Parmer PTFE Beaker Cover for 06300-78
Cole-Parmer | Item: 0630089
Stand up to the harshest chemicals
₹3,200.00INR / Each
₹3,200.00INR / Each
Product Description
Key Features
  • Constructed of chemically inert PTFE
  • Autoclavable
  • Beaker cover is suitable for 150 mL beaker 06300-78
More About this Item
These PTFE beaker covers are compatible with the 06300-series PTFE broad spout beakers.
Specifications
  • MaterialPolytetrafluoroethylene (PTFE)
  • Cap MaterialPolytetrafluoroethylene (PTFE)
  • Cap ColorWhite
  • AutoclavableYes
  • Withstands High Temperature (DegreeF)572
  • Withstands Low Temperature (° F)-328
  • Outside Diameter (OD) (cm)7.5
  • DescriptionPTFE Beaker Cover for 06300-78, 1/ea