Cole-Parmer PTFE Beaker Cover for 06300-74
Representative image only.
Cole-Parmer PTFE Beaker Cover for 06300-74
Cole-Parmer | Item: 0630085
Stand up to the harshest chemicals
₹2,600.00INR / Each
₹2,600.00INR / Each
Product Description
Key Features
  • Constructed of chemically inert PTFE
  • Autoclavable
  • Beaker cover is suitable for 50 mL beaker 06300-74
More About this Item
These PTFE beaker covers are compatible with the 06300-series PTFE broad spout beakers.
Specifications
  • MaterialPolytetrafluoroethylene (PTFE)
  • Cap MaterialPolytetrafluoroethylene (PTFE)
  • Cap ColorWhite
  • AutoclavableYes
  • Withstands High Temperature (DegreeF)572
  • Withstands Low Temperature (° F)-328
  • Outside Diameter (OD) (cm)5
  • DescriptionPTFE Beaker Cover for 06300-74, 1/ea