Cole-Parmer Pressure Sensor, Nonsterile, Polycarbonate, Luer Connections
Representative image only.
Cole-Parmer Pressure Sensor, Nonsterile, Polycarbonate, Luer Connections
Cole-Parmer | Item: 4852310
Measure the static and dynamic pressure of gases and liquids accurately up to 75 psi
₹12,400.00INR / Each
₹12,400.00INR / Each
Product Description
Includes
12" white cable with over-molded 4-position-UTMD connector and a certificate of compliance.
Key Features
  • High-pressure piezo-resistive transducer provides highly accurate pressure sensing
  • Flow-through design reduces dead space and pressure drop
  • Save money with single-use pressure sensor
  • Manufactured in ISO 13485 certified facilities in ISO Class 7 cleanrooms
  • All fluid contact components meet USP Class VI and ISO 10993-5 requirements
  • Latex free and animal-derivative component free
  • Sterilize by gamma irradiation or ethylene oxide
  • Polycarbonate construction
  • Luer connections
More About this Item
The single-use pressure sensors feature a high-pressure piezo-resistive transducer to accurately measure static and dynamic pressures of gases and liquids up to 75 psi. Unobstructed, flow-through design minimizes dead space and pressure drops. Use in multi-stage depth filtration, TFF/crossflow filtration, chromatography processes, glass and disposable bioreactor pressure monitoring, and filling operations applications. Pressure sensors come with Luer fittings for quick, leak-free connections.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Accuracy-10 to -7psi ±5% of reading,-7 to 0psi ±3% of reading,0 to 6psi ±2% of reading,6 to 30psi ±3% of reading,30 to 60psi ±5% of reading
  • Min Pressure (PSI)-11.5
  • Max Pressure (PSI)75
  • Min Temperature (° F)35.6
  • Max Temperature (° F)104
  • Min Temperature (° C)2
  • Max Temperature (DegreeC)40
  • Wetted MaterialsPolycarbonate
  • Process ConnectionLuer
  • Output0.2584 μV/V/PSI
  • Media CompatibilityAll materials in contact with fluid path meet USP Class VI
  • SterileNo
  • DescriptionPressure Sensor, Nonsterile, Polycarbonate, Luer Connections