Cole-Parmer Prepared Microscope Slide Set, Biology, Set of 12 Slides; 1/EA
Representative image only.
Cole-Parmer Prepared Microscope Slide Set, Biology, Set of 12 Slides; 1/EA
Cole-Parmer | Item: 7890848
Convenient collection of prepared slides
₹13,800.00INR / Each
₹13,800.00INR / Each
Product Description
Includes
Glass prepared microscope slides supplied in a plastic case.
Key Features
  • Numbered indexes allow for easy identification
  • Microscope slides are constructed of durable glass
More About this Item
These Cole-Parmer® Essentials prepared microscope slide sets include a wide assortment of specimen slides. All prepared slide collections come packaged in a case for easy and safe storage.
A comprehensive selection of prepared microscope slides, the set includes prepared slides depicting:
Bacteria (smear from mouth)
Onion root tip (mitosis longitudinal section)
Amoeba Proteus (whole mount)
Monocot/Dicot Stem (cross-section)
Paramecium (whole mount)
Lilac leaf (cross-section)
Spirogyra vegetative filaments (whole mount)
Hydra (whole mount)
Bread Molds Sexual (whole mount)
Earthworm (cross-section of posterior)
Buttercup Root (cross-section)
Human Blood Smear
Specifications
  • Slide TypeBiology, Set of 12
  • DescriptionPrepared Microscope Slide Set, Biology, Set of 12 Slides; 1/EA