Cole-Parmer Port Disk Threaded Fitting, Natural Kynar® Flex, Cleanroom Packed, 38-400 1" Thread; 10/Pk
Representative image only.
Cole-Parmer Port Disk Threaded Fitting, Natural Kynar® Flex, Cleanroom Packed, 38-400 1" Thread; 10/Pk
Cole-Parmer | Item: 5011689
Weld to single-use bags to transfer viscose fluids and powders
₹16,900.00INR / Pkg of 10
₹16,900.00INR / Pkg of 10
Product Description
Key Features
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Wide 1" opening—ideal for powders and high viscosity fluid
  • Ultrasonic, radio frequency, or thermal weld to Kynar bags
  • 38-400 1" Thread
  • Natural Kynar® Flex
  • Meets FDA requirements 21 CFR 177.1520(c), 21 CFR 176.170(c), 21 CFR 177.2600(c)(4)(i), NSF/ANSI Standard 51 and 61, and RoHS compliant
More About this Item
Use these high-quality PVC-free port disks with 38-400 bottle cap thread and 1" wide opening to transfer high viscosity fluids and powders from single-use bags used in pharmaceutical and medical industries. Ultrasonic, radio frequency, or thermal weld port disks to polyethylene or Kynar medical and bioreactor bags to easily transfer fluids.
Disks are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Natural Kynar® fittings provide excellent chemical, flame, gamma radiation, UV, and weather resistance. These robust fittings exhibit good mechanical properties, low moisture absorption, and good machinability. Natural Kynar®, also known as natural Polyvinylidene Fluoride (PVDF), is a PFAS-free resin.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPVDF
  • Sterilization MethodGamma irradiation between 25 and 100 kGy, EtO, or autoclave at 15 psi steam at 121°C (250°F)
  • Connection TypePort Disk x Thread
  • Flow PatternStraight
  • ColorNatural
  • Regulatory ApprovalsNon-Phthalate;FDA;Non-DEHP;NSF-61;NSF-51;RoHS
  • DescriptionPort Disk Threaded Fitting, Natural Kynar® Flex, Cleanroom Packed, 38-400 1" Thread; 10/Pk