Cole-Parmer Port Disk Hose Barb Fitting, Engage™ 8402, No Dimples, Cleanroom Packed, 1/8" ID; 10/Pk
Representative image only.
Cole-Parmer Port Disk Hose Barb Fitting, Engage™ 8402, No Dimples, Cleanroom Packed, 1/8" ID; 10/Pk
Cole-Parmer | Item: 5011705
Weld to single-use bags to transfer cell culture, drug compounds, and other biological fluids
₹3,700.00INR / Pkg of 10
₹3,700.00INR / Pkg of 10
Product Description
Key Features
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Flexible, smooth flat base for easy attachment
  • Ultrasonic, radio frequency, or thermal weld to polyethylene bags
  • Meets FDA requirements 21 CFR 177.1520(c) 3.2c and 21 CFR 176.170(c) and RoHS compliant
  • 1/8" hose barb
More About this Item
Use these high-quality PVC-free port disks with hose barb to transfer fluids, buffers, media, and drug compounds from single-use bags used in pharmaceutical and medical industries. Ultrasonic, radio frequency, or thermal weld port disks to polyethylene medical and bioreactor bags to easily transfer fluids.
Engage™ 8402 polyolefin material is odorless, translucent, and animal-derivative-free ethylene-octene copolymer that offers high clarity for easier visual inspection. It offers excellent durability while providing flexibility. The flexible disk curves with the bag and the barb flexes, minimizing the risk of it cracking with sideload. The polymer withstands aqueous acids, bases, alcohols, glycols, esters, and ketones.
Disks are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialEngage™ 8402
  • Sterilization MethodGamma irradiation up to 50 kGy
  • Connection TypePort Disk x Hose Barb
  • Barb Size / Tubing Id (Port 1) (in)1/8
  • Flow PatternStraight
  • ColorTranslucent
  • Regulatory ApprovalsADCF;FDA;RoHS;USP;Non-DEHP;Non-Phthalate
  • DescriptionPort Disk Hose Barb Fitting, Engage™ 8402, No Dimples, Cleanroom Packed, 1/8" ID; 10/Pk