Cole-Parmer PolarSafe® Cryogenic Storage and Transport Dewar, 35L with Smart Cap; 1/Ea
Representative image only.
Cole-Parmer PolarSafe® Cryogenic Storage and Transport Dewar, 35L with Smart Cap; 1/Ea
Cole-Parmer | Item: 1634033
Robust cryopreservation solutions for long-term storage and transport of biological samples
₹272,000.00INR / Each
₹272,000.00INR / Each
Product Description
Key Features
  • Strong yet lightweight aluminum dewars hold liquid nitrogen
  • Narrow neck design provides high thermal efficiency and low evaporation loss
  • Long term storage up to 76 days
  • 5 year vacuum warranty
  • 1 year warranty
More About this Item
Confidently store your samples in liquid nitrogen, with ulta-low evaporation loss. With two handles, these dewars are easy to transport. Each unit has numbered index location points for canisters, and can also accommodate straw storage. Long-term storage duration under normal operating conditions.
Smart cap includes enhance storage and monitoring your temperature sensitive materials.
1 Year WarrantyConformite Europeenne
Specifications
  • DescriptionCryo Dewar, 35 L with Racks with Smart Cap; 1/Ea