Cole-Parmer PolarSafe® Cryogenic Storage and Transport Dewar, 175 L; 1/Ea
Representative image only.
Cole-Parmer PolarSafe® Cryogenic Storage and Transport Dewar, 175 L; 1/Ea
Cole-Parmer | Item: 1634037
Robust cryopreservation solutions for long-term storage and transport of biological samples
₹545,000.00INR / Each
₹545,000.00INR / Each
Product Description
Key Features
  • Strong yet lightweight aluminum dewars hold liquid nitrogen
  • Narrow neck design provides high thermal efficiency and low evaporation loss
  • Long term storage up to 184 days
  • 5 year vacuum warranty
  • 1 year warranty
More About this Item
Confidently store your samples in liquid nitrogen, with ultra-low evaporation loss. With one or two handles depending on model, these dewars are easy to transport. Each unit has numbered index location points for canisters, and can also accommodate straw storage. Long-term storage duration varies by model but can last up to 200 days under normal operating conditions.
Order lockable cap separately. Enhance monitoring and ease of transportation with these Cole-Parmer® Polarsafe® Dewar Accessories which includes Smart Caps, trolleys, dewar monitor, and level rulers.
1 Year WarrantyConformite Europeenne
Specifications
  • DescriptionLiquid Nitrogen Container, Aluminium, 175 L; 1/Ea