Cole-Parmer Plastic Pasteur Pipettes, 3 mL, Bulk, Individually Wrapped; 300/CS
Representative image only.
Cole-Parmer Plastic Pasteur Pipettes, 3 mL, Bulk, Individually Wrapped; 300/CS
Cole-Parmer | Item: 1635173
Easily draw and deliver precise solutions
₹3,300.00INR / Case of 300
₹3,300.00INR / Case of 300
Product Description
Key Features
  • Deliver repeatable, uniform drops
  • Eliminate cross contamination with these low cost, disposable LDPE pipettes
  • 1 year warranty
More About this Item
Use these unbreakable low-density polyethylene (LDPE) pipettes to eliminate the chance of contamination from rubber bulbs. They deliver repeatable, uniform drops and are configured to fit into a variety of vessels. Choose the bulb draw and graduations or drops/mL needed for measurement and delivery.
1 Year Warranty
Specifications
  • SterileNo
  • Autoclavableyes
  • Max Volume (µL)3000
  • Max Volume (mL)3
  • Package Of300/Cs
  • DescriptionCole-Parmer Plastic Pasteur Pipettes, 3 mL, Bulk, Individually Wrapped; 300/Cs