Cole-Parmer Plastic Pasteur Pipettes, 1 mL, Bulk, Individually Wrapped; 3000/CS
Representative image only.
Cole-Parmer Plastic Pasteur Pipettes, 1 mL, Bulk, Individually Wrapped; 3000/CS
Cole-Parmer | Item: 1597824
Easily draw and deliver precise solutions
₹11,100.00INR / Case of 3000
₹11,100.00INR / Case of 3000
Product Description
Key Features
  • Deliver repeatable, uniform drops
  • Eliminate cross contamination with these low cost, disposable LDPE pipettes
  • 1 year warranty
More About this Item
Use these unbreakable low-density polyethylene (LDPE) pipettes to eliminate the chance of contamination from rubber bulbs. They deliver repeatable, uniform drops and are configured to fit into a variety of vessels. Choose the bulb draw and graduations or drops/mL needed for measurement and delivery.
1 Year Warranty
Specifications
  • SterileNo
  • Max Volume (µL)1000
  • Autoclavableyes
  • Max Volume (mL)1
  • Package Of3000/Cs
  • DescriptionCole-Parmer Plastic Pasteur Pipettes, 1 mL, Bulk, Individually Wrapped; 3000/Cs