Cole-Parmer Pipettor Rack, 8-Place with 6 Multichannel Slots
Representative image only.
Cole-Parmer Pipettor Rack, 8-Place with 6 Multichannel Slots
Cole-Parmer | Item: 1400311
Unique design accommodates both multichannel and standard pipettes
₹33,600.00INR / Each
₹33,600.00INR / Each
Buy It Together
Current Item
Cole-Parmer Pipettor Rack, 8-Place with 6 Multichannel Slots
Cole-Parmer Pipettor Rack, 8-Place with 6 Multichannel Slots
1400311
₹33,600.00INR / Each
+
Cole-Parmer Adjustable-Volume Pipettor, 500 to 5000 µL
2450109
₹8,400.00INR / Each
=
Total Price 42000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • The patented open slot allow multichannel head to fit on the rack
  • 5-mm thick acrylic offers durability along with stability
  • Four nonskid rubber feet provide extra stability
  • Accepts eight pipettors, up to six multichannel pipettors
More About this Item
Store single-channel pipettors along with multichannel pipettors on a single pipette rack. A unique design featuring a slot accommodates a multichannel head longways through the rack. Pipettors rest securely in place at two places, under the finger hook and near the bottom of the handle, for added stability.
Specifications
  • MaterialAcrylic
  • ColorWhite
  • Compatible With8 pipettors, up to 6 multichannel pipettors
  • DescriptionSureStand™ XL, Pipettor Rack, 8-Place with 6 Multichannels