Cole-Parmer Pipettor Rack, 5-Place with 2 Multichannel Slots
Representative image only.
Cole-Parmer Pipettor Rack, 5-Place with 2 Multichannel Slots
Cole-Parmer | Item: 1400309
Unique design accommodates both multichannel and standard pipettes
₹25,000.00INR / Each
₹25,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer Pipettor Rack, 5-Place with 2 Multichannel Slots
Cole-Parmer Pipettor Rack, 5-Place with 2 Multichannel Slots
1400309
₹25,000.00INR / Each
+
Cole-Parmer Adjustable-Volume Pipettor, 500 to 5000 µL
2450109
₹8,400.00INR / Each
=
Total Price 33400.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • The patented open slot allow multichannel head to fit on the rack
  • 5-mm thick acrylic offers durability along with stability
  • Four nonskid rubber feet provide extra stability
  • Accepts five pipettors, up to two multichannel pipettors
More About this Item
Store single-channel pipettors along with multichannel pipettors on a single pipette rack. A unique design featuring a slot accommodates a multichannel head longways through the rack. Pipettors rest securely in place at two places, under the finger hook and near the bottom of the handle, for added stability.
Specifications
  • MaterialAcrylic
  • ColorWhite
  • Compatible With5 pipettors, up to 2 multichannel pipettors
  • DescriptionSureStand™ Pipettor Rack, 5-Place with 2 Multichannels