Cole-Parmer Pipette Rack, 100 to 1000 µL Long Tip; 6/PK
Representative image only.
Cole-Parmer Pipette Rack, 100 to 1000 µL Long Tip; 6/PK
Cole-Parmer | Item: 2450308
Stackable color-coded trays to house your tips in a safe and contaminate-free environment
₹5,500.00INR / Pkg of 6
₹5,500.00INR / Pkg of 6
Product Description
Key Features
  • Fully autoclavable and stackable
  • Color-coded tray for easy identification
  • Hinge rack allows single hand operation
  • Free from DNase, RNase, Human DNA, Pyrogen and PCR inhibitors
  • Holds 100 to 1000 µL long tips
More About this Item
When it comes to reaching for that specific pipette tip, these racks make is easy. They can hold standard and long tips with various volume measurements and, are available in packages of 6 or 10 trays.
Specifications
  • AutoclavableYes
  • Max Volume (µL)1000
  • Min Volume (µL)100
  • Tip StyleLong
  • Color CodeYes
  • Package Of6 racks
  • Compatible withLong 9.82 cm tips
  • DescriptionOmega Pipette Rack for 100 to 1000 µL long tips, 6/PK