Cole-Parmer pH Electrode Housing, submersible, PVDF w/ Viton o-rings
Representative image only.
Cole-Parmer pH Electrode Housing, submersible, PVDF w/ Viton o-rings
Cole-Parmer | Item: 0599393
Protective housings for 12-mm electrodes
₹61,700.00INR / Each
₹61,700.00INR / Each
Product Description
Key Features
  • Protective housings provide an economical alternative
More About this Item
This 41''L x 1-1/16'' OD pipe is welded to a PVC connection box, and sealed with VITON O-rings for superior protection. Bottom tube detaches for electrode installation. CPVC model withstands temperatures up to 176°>F (80°>C); KYNAR model up to 230°>F (110°>C). Order extension cables for both models separately.
Specifications
  • ApplicationsSubmersible
  • ConnectorBNC
  • DescriptionpH Electrode Housing, submersible, PVDF w/ Viton o-rings