Cole-Parmer pH electrode, combination, rugged, epoxy body, interchangeable connection
Representative image only.
Cole-Parmer pH electrode, combination, rugged, epoxy body, interchangeable connection
Cole-Parmer | Item: 0565845
Save space and money by using one cable with multiple electrodes or vice versa
₹15,000.00INR / Each
₹15,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer pH electrode, combination, rugged, epoxy body, interchangeable connection
Cole-Parmer pH electrode, combination, rugged, epoxy body, interchangeable connection
0565845
₹15,000.00INR / Each
+ =
Total Price 25000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Cable cap rotates independently and will not bunch up
  • 6 month warranty
More About this Item
Match these combination electrodes with one of the connector cables listed at the right. If you use several pH meters that each have different connector types, these electrodes and cables can save you money. The probe-cable interface is moisture proof and noise free, with a sealing O-ring, gold-plated contacts, and PTFE insulation.
6 Month Warranty
Specifications
  • Electrode TypeSingle junction
  • Electrode Body MaterialEpoxy
  • Reference ElectrolyteAg/AgCl
  • Max Temperature (° C)80
  • ConnectorDepends upon cable purchased separately: BNC, US standard, or SN6 to 57
  • Diameter (mm)12
  • Length (mm)127
  • DescriptionpH electrode, combination, rugged, epoxy body, interchangeable connection
  • Warranty6 month warranty