Cole-Parmer Petri Dish Seals; 400/Pk
Representative image only.
Cole-Parmer Petri Dish Seals; 400/Pk
Cole-Parmer | Item: 7595509
No more wrapping dishes!
₹70,800.00INR / Pkg of 400
₹70,800.00INR / Pkg of 400
Product Description
Key Features
  • Block evaporation during extended incubation times
  • Extend sensitive, pre-made plates' shelf life
  • Contain pathogenic microorganisms to increase safety
  • Compatible with standard 100-mm diameter plates
More About this Item
These seals are an effective and efficient alternative to film wrapping your plates and dishes. Seals apply in seconds. Slip the moist band over the dish's edge—no gaps, no ripping!
Specifications
  • ColorTranslucent
  • MaterialCellulose acetate
  • Width (cm)9.2
  • DescriptionPetri Dish Shrink Seals; 400/Pk