Cole-Parmer PES Sterile Chromatography Syringe Filters; 0.45µm Pore, 25mm Dia, 50/Pk

Representative image only.

Cole-Parmer PES Sterile Chromatography Syringe Filters; 0.45µm Pore, 25mm Dia, 50/Pk

Cole-Parmer | Item: 1594553

₹4,700.00INR / Each

₹4,700.00INR / Each
Product Description

Key Features

  • Bidirectional flow with female luer lock on one side and a male luer slip on the other
  • Withstand pressure up to 100 psi (7 bar)
  • Batch test certificate included with each pack

More About this Item

PES Syringe Filters are naturally low protein binding and sample absorbing, and have a wide chemical compatibility for both aqueous and organic solvent filtration. Recommended for filtering critical biological sampling, tissue culture media, additives, serum and buffers. Maximum use temperature is 50°C (122°F).

Sterile
Specifications
  • Membrane MaterialPolyethersulfone (PES)
  • Pore Size (µm)0.45
  • Membrane Diameter (mm)25
  • Housing MaterialPolypropylene (PP)
  • Sample Volume (mL)10 to 100 mL
  • SterileYes
  • ConnectionsMale Slip Tip Luer/Female Luer Lock
  • Retention Volume (µL)<50
  • Effective Filtration Area (cm²)3.9
  • Max Pressure (PSI)100
  • Max Pressure (bar)7
  • Max Operating Temperature (° F)122
  • Max Operating Temperature (° C)50
  • DescriptionPES Sterile Chromatography Syringe Filters; 0.45µm Pore, 25mm Dia, 50/Pk
Reviews
Q&A