Cole-Parmer PCR-300-S Thermal Cycler, Single 384-Well Capacity; 100 to 230 VAC

Representative image only.

Cole-Parmer PCR-300-S Thermal Cycler, Single 384-Well Capacity; 100 to 230 VAC

Cole-Parmer | Item: 9394500

Easily create, modify and access programs, saving time

Call For Price

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

Call For Price

Buy It Together

Current Item

Product Description

Key Features

  • Flexible—your choice of one, two, or four block capacities in 96-well, 384-well, or mixed formats
  • Independently control each block for simplified temperature and space optimization
  • Software allows quick access to commonly used protocols
  • Single block system features 384-well capacity
  • 3 year warranty

More About this Item

These compact thermal cyclers deliver reproducible results every run—with little to no training needed. Features include a clear responsive touch screen, secure user-specific programming, adjustable heated lid, and active sample cooling for sharper amplification.

A Program Wizard generates a protocol specific to your sequence in seconds—a quick way to optimize new assays. Units also retain approximately 1000 reports for reviewing at a later date and run on an intuitive, HD Android™ tablet interface. Save your personal settings to your USB drive; sign in is as simple as inserting your USB drive, no password needed.

3 Year WarrantyConformite Europeenne
Specifications
  • Max Temperature (° C)100
  • Min Temperature (° C)10
  • Temperature Accuracy50⁰C: ±0.25⁰C
  • Max Gradient (° C)29
  • Min Gradient (° C)1
  • Max Heating Rate3.4⁰C per second
  • Heated Lid Temperature35 to 115⁰C, or off
  • Heated Lid PressureAdjustable
  • Number of Blocks1
  • Block UniformityBlock at 55⁰C: ±0.3⁰C
  • Block DescriptionOne 384-well plate
  • Memory1000 programs max
  • OutputUSB port
  • Auto Restart On Power FailureYes
  • Height (in)10 1/4
  • Width (in)16 15/16
  • Depth (in)13
  • Height (cm)26
  • Width (cm)43
  • Depth (cm)33
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • DescriptionThermal Cycler, Single 384-Well Capacity; 100-230 VAC
  • Warranty3 Years
Reviews
Q&A