Cole-Parmer Paperless Recorder, 2-Channel; Univ AC Adapter/Rechargeable Battery
Representative image only.
Cole-Parmer Paperless Recorder, 2-Channel; Univ AC Adapter/Rechargeable Battery
Cole-Parmer | Item: 8081640
No more paper to jam, no more pens to clog
Call For Price
for an additional
₹43,800.00INR / Each
for an additional
₹56,400.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
Call For Price
Buy It Together
Current Item
Cole-Parmer Paperless Recorder, 2-Channel; Univ AC Adapter/Rechargeable Battery
Cole-Parmer Paperless Recorder, 2-Channel; Univ AC Adapter/Rechargeable Battery
8081640
Call For Price
+ + =
Total Price 96500.00 ADD 3 ITEMS TO CART
Product Description
Includes
1 rechargeable lithium-ion battery.
Key Features
  • Intuitive operation, flexible input configurations, and large memory capabilities
  • Highly reflective LCD with bright white LED backlight displays real-time data in digital or trend format
  • Two universally configurable isolated inputs for measuring DC voltage, DC current, thermocouples, RTDs, as well as frequency and pulse inputs
  • Four internal alarm set points, two alarm relay outputs, and one digital control input
  • Ethernet and USB communication options available
  • 1 year warranty
More About this Item
Compact size combined with extreme flexibility makes these two-channel paperless recorders a powerful, yet economical, digital data recording system. A maximum storage rate of 100 samples per second can be set for both channels, letting you capture short duration process signals anomalies. Use CompactFlash™ cards up to 2 GB for data storage over longer time periods.
Conformite Europeenne1 Year Warranty
Specifications
  • Number of Channels2
  • Chart TypeDigital
  • Max Temperature (° F)4350
  • Min Temperature (° F)-150
  • Max Temperature (° C)2400
  • Min Temperature (° C)-101
  • Battery Type1 rechargeable lithium-ion (included)
  • DescriptionPaperless Recorder, 2-Channel; Univ AC Adapter/Rechargeable Battery