Cole-Parmer Ornithobacterium rhinotracheale, Without Mastermix
Representative image only.
Cole-Parmer Ornithobacterium rhinotracheale, Without Mastermix
Cole-Parmer | Item: 9394734
Quickly and accurately detect the presence of avian infections
₹206,000.00INR / Each
₹206,000.00INR / Each
Product Description
Includes
Lyophilized primer and probe mix, positive control for standard curve, internal extraction control DNA, endogenous control, and RNAse/DNAse-free water.
Key Features
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
More About this Item
Each kit is designed to ensure the broadest possible detection profile and detection of all relevant strains and subtypes. Order a complete kit with mastermix or a kit without mastermix and use your own mastermix.
Specifications
  • Test TypeDNA
  • Number of Tests Included150
  • Mastermix IncludedNo
  • Binomial NameOrnithobacterium rhinotracheale
  • DescriptionOrnithobacterium rhinotracheale, Without Mastermix