Cole-Parmer OmegaZen™ Replacement Nosepiece
Representative image only.
Cole-Parmer OmegaZen™ Replacement Nosepiece
Cole-Parmer | Item: 2530354
Enhance and maintain your Omega Zen pipettors with these accessories
₹1,200.00INR / Each
₹1,200.00INR / Each
Product Description
Key Features
  • Compatible with Omega Zen pipette controllers
  • Choose from a wide range of accessories to make your pipettor even more versatile
  • Autoclavable
  • 1 year warranty
More About this Item
These accessories can be used to enhance the flexibility of your Omega Zen controller, or be used for routine maintenance. There are four filter options to choose from. All are made of a polypropylene housing with a hydrophobic PTFE membrane. The Omega Zen controllers accept either the 0.2 µm or the 0.45 µm pore size filters.
Power your Omega Zen controller with the rechargeable lithium-ion battery for up to 9 hours of continuous use and charge the battery with the universal power supply.
The nosepiece and silicone pipette adapter are both autoclavable for optional sterility.
1 Year Warranty
Specifications
  • SterileNo
  • AutoclavableYes
  • Body MaterialUV-resistant plastic
  • Compatible withArgos Technologies OmegaZen Pipette Controller
  • DescriptionReplacement Nosepiece