Cole-Parmer OmegaZen™ Filters, 0.45 µm, Sterile; 10/Pk
Representative image only.
Cole-Parmer OmegaZen™ Filters, 0.45 µm, Sterile; 10/Pk
Cole-Parmer | Item: 2530339
Enhance and maintain your Omega Zen pipettors with these accessories
₹5,300.00INR / Pkg of 10
₹5,300.00INR / Pkg of 10
Product Description
Key Features
  • Compatible with Omega Zen pipette controllers
  • Choose from a wide range of accessories to make your pipettor even more versatile
  • 1 year warranty
More About this Item
These accessories can be used to enhance the flexibility of your Omega Zen controller, or be used for routine maintenance. There are four filter options to choose from. All are made of a polypropylene housing with a hydrophobic PTFE membrane. The Omega Zen controllers accept either the 0.2 µm or the 0.45 µm pore size filters.
Power your Omega Zen controller with the rechargeable lithium-ion battery for up to 9 hours of continuous use and charge the battery with the universal power supply.
The nosepiece and silicone pipette adapter are both autoclavable for optional sterility.
The filter should be replaced every three months or when the filter is contaminated by liquid or the suction capacity is slowed.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
1 Year Warranty
Specifications
  • SterileYes
  • Body MaterialHydrophobic PTFE
  • Filter Type0.45 µm
  • Compatible withArgos Technologies OmegaZen Pipette Controller
  • DescriptionFilters, 0.45 µm, Sterile; 10/pk