Cole-Parmer NS-SHI-101 Needle Seal, Shimadzu SIL-10; 1/EA
Representative image only.
Cole-Parmer NS-SHI-101 Needle Seal, Shimadzu SIL-10; 1/EA
Cole-Parmer | Item: 1195416
Keep your instruments running smoothly with these replacement parts
₹21,800.00INR / Each
₹21,800.00INR / Each
Product Description
Key Features
  • Compatible with Shimadzu chromatography instruments
  • OEM reference number: 228-33355-04
More About this Item
Optimize performance of your Shimadzu chromatography instruments with these replacement parts.
Specifications
  • Compatible WithShimadzu SIL-10ADvp, SIL-10AXL
  • DescriptionNeedle Seal, Shimadzu SIL-10; 1/EA