Cole-Parmer NGDI-20 Monobed Deionizer Mixed-Bed Resin Cartridge; Ultrapure
Representative image only.
Cole-Parmer NGDI-20 Monobed Deionizer Mixed-Bed Resin Cartridge; Ultrapure
Cole-Parmer | Item: 0150312
Keep your monbed deionizer running smoothly
₹20,500.00INR / Each
₹20,500.00INR / Each
Product Description
Key Features
  • Designed to be easily interchanged with your monobed deionizer
  • Ideal for removing fine particulates
  • 1 year warranty
More About this Item
Choose from resins and an ultrapure cartidge to provide high purity water from your monobed deionizer water systems.
To install the resin cartridge, remove the tube from inside the deionizer. Install the cartridge with screen end down and the single hole end up. When using a cartridge with housing 01503-00, the tube included with the housing must be removed. Do NOT remove the plugs in the cartridge (red and black plugs).
1 Year Warranty
Specifications
  • Compatible WithMonobed Deionizer System
  • DescriptionMonobed Deionizer Mixed-Bed Resin Cartridge; Ultrapure