Cole-Parmer Nester Extension Clamp; Nickel-Plated Zinc and Ss
Representative image only.
Cole-Parmer Nester Extension Clamp; Nickel-Plated Zinc and Ss
Cole-Parmer | Item: 0802350
Provide support for your specific laboratory equipment
₹17,900.00INR / Each
₹17,900.00INR / Each
Product Description
Key Features
  • Durable stainless steel clamps offer chemical resistance and increased strength
  • NOTE For most Specially and Support Clamps, an additional holder must be purchased in order to attach clamps to lab-frames, support stands or other apparatus
  • 1 year warranty
More About this Item
Securely holds your glass distillation columns, odd-shaped glassware and most other labware. NOTE: An additional connector must be purchased in order to attach clamps to frames or other apparatus.
1 Year Warranty
Specifications
  • Clamp MaterialNickel-plated Zinc
  • Rod MaterialStainless Steel
  • Clamp TypeEquipment, Nester Extension
  • Min Grip Range (InchShort)1.97
  • Max Grip Range (InchShort)4
  • Length (in)10.32
  • Depth (in)0.39
  • Diameter (in)0.39
  • DescriptionNester Extension Clamp; Nickel-Plated Zinc and Ss