Cole-Parmer Narrow-Mouth Transport Plastic Bottle, HDPE, 4000mL

Representative image only.

Cole-Parmer Narrow-Mouth Transport Plastic Bottle, HDPE, 4000mL

Cole-Parmer | Item: 0625360

Transport hazardous samples reliably

₹5,900.00INR / Pkg of 1

₹5,900.00INR / Pkg of 1

Buy It Together

Current Item

Cole-Parmer Narrow-Mouth Transport Plastic Bottle, HDPE, 4000mL
Cole-Parmer Narrow-Mouth Transport Plastic Bottle, HDPE, 4000mL
0625360

₹5,900.00INR / Pkg of 1
+
Cole-Parmer Replacement Cap, PP, 38-430; 12/PK
6300018

₹5,900.00INR / Pkg of 12
=
Total Price 11800.00 ADD 2 ITEMS TO CART
Product Description

Includes

Polypropylene screw closures.

Key Features

  • Safe for small quantities of corrosives or cleaning supplies
  • Leakproof, PP screw cap
  • 1 year warranty

More About this Item

Ideal for small-scale inter-lab shipping of hazardous samples and chemicals such as corrosives or cleaning compounds.

Certificate of Analysis
For a sample Certificate of Analysis, please go to link below under RESOURCES.
If a Lot specific Certificate of Analysis is required, please call 1-800-323-4340 for assistance.

1 Year Warranty
Specifications
  • Capacity (mL)4000
  • Capacity (L)4
  • Capacity (oz)128
  • Capacity (Gal)1.05680317040951
  • MaterialHigh Density Polyethyene (HDPE)
  • Cap MaterialPolypropylene
  • Cap / Stopper Size38-430
  • Cap Colornatural
  • Bottle Colornatural
  • AutoclavableNo
  • Neck Inside Diameter (ID) (in)0
  • Outside Diameter (OD) (in)6 1/16
  • Outside Diameter (OD) (cm)15.4
  • Overall Height (in)12 51/64
  • Overall Height (cm)32.5
  • Overall Depth (in)6 1/16
  • Overall Depth (cm)15.4
  • Overall Width (in)6 1/16
  • Overall Width (cm)15.4
  • DescriptionNarrow-Mouth Transport Plastic Bottle, HDPE, 4000mL
  • Warranty1 year
Reviews
Q&A