Cole-Parmer Narrow-Mouth Transport Plastic Bottle, HDPE, 250mL; 12/PK

Representative image only.

Cole-Parmer Narrow-Mouth Transport Plastic Bottle, HDPE, 250mL; 12/PK

Cole-Parmer | Item: 0625330

Transport hazardous samples reliably

₹7,500.00INR / Pkg of 12

₹7,500.00INR / Pkg of 12
Product Description

Includes

Polypropylene screw closures.

Key Features

  • Safe for small quantities of corrosives or cleaning supplies
  • Leakproof, PP screw cap
  • 1 year warranty

More About this Item

Ideal for small-scale inter-lab shipping of hazardous samples and chemicals such as corrosives or cleaning compounds.

Certificate of Analysis
For a sample Certificate of Analysis, please go to link below under RESOURCES.
If a Lot specific Certificate of Analysis is required, please call 1-800-323-4340 for assistance.

1 Year Warranty
Specifications
  • Capacity (mL)250
  • Capacity (L)0.25
  • Capacity (oz)8
  • Capacity (Gal)0.0660501981505945
  • MaterialHigh Density Polyethyene (HDPE)
  • Cap MaterialPolypropylene
  • Cap / Stopper Size24
  • Cap Colornatural
  • Bottle Colornatural
  • AutoclavableNo
  • Neck Inside Diameter (ID) (mm)17.3
  • Neck Inside Diameter (ID) (in)11/16
  • Outside Diameter (OD) (in)2 2/5
  • Outside Diameter (OD) (cm)6.1
  • Overall Height (in)5 1/8
  • Overall Height (cm)13
  • Overall Depth (in)2 2/5
  • Overall Depth (cm)6.1
  • Overall Width (in)2 2/5
  • Overall Width (cm)6.1
  • DescriptionNarrow-Mouth Transport Plastic Bottle, HDPE, 250mL; 12/PK
  • Warranty1 year
Reviews
Q&A