Cole-Parmer Narrow-Mouth Transport Plastic Bottle, HDPE, 1000mL; 6/PK

Representative image only.

Cole-Parmer Narrow-Mouth Transport Plastic Bottle, HDPE, 1000mL; 6/PK

Cole-Parmer | Item: 0625350

Transport hazardous samples reliably

₹7,600.00INR / Pkg of 6

₹7,600.00INR / Pkg of 6

Buy It Together

Current Item

Cole-Parmer Narrow-Mouth Transport Plastic Bottle, HDPE, 1000mL; 6/PK
Cole-Parmer Narrow-Mouth Transport Plastic Bottle, HDPE, 1000mL; 6/PK
0625350

₹7,600.00INR / Pkg of 6
+
Cole-Parmer Replacement Cap, PP, 38-430; 12/PK
6300018

₹5,900.00INR / Pkg of 12
=
Total Price 13500.00 ADD 2 ITEMS TO CART
Product Description

Includes

Polypropylene screw closures.

Key Features

  • Safe for small quantities of corrosives or cleaning supplies
  • Leakproof, PP screw cap
  • 1 year warranty

More About this Item

Ideal for small-scale inter-lab shipping of hazardous samples and chemicals such as corrosives or cleaning compounds.

Certificate of Analysis
For a sample Certificate of Analysis, please go to link below under RESOURCES.
If a Lot specific Certificate of Analysis is required, please call 1-800-323-4340 for assistance.

1 Year Warranty
Specifications
  • Capacity (mL)1000
  • Capacity (L)1
  • Capacity (oz)32
  • Capacity (Gal)0.264200792602378
  • MaterialHigh Density Polyethyene (HDPE)
  • Cap MaterialPolypropylene
  • Cap / Stopper Size38-430
  • Cap Colornatural
  • Bottle Colornatural
  • AutoclavableNo
  • Neck Inside Diameter (ID) (mm)27.3
  • Neck Inside Diameter (ID) (in)1 5/64
  • Outside Diameter (OD) (in)3 35/64
  • Outside Diameter (OD) (cm)9
  • Overall Height (in)8 17/64
  • Overall Height (cm)21
  • Overall Depth (in)3 35/64
  • Overall Depth (cm)9
  • Overall Width (in)3 35/64
  • Overall Width (cm)9
  • DescriptionNarrow-Mouth Transport Plastic Bottle, HDPE, 1000mL; 6/PK
  • Warranty1 year
Reviews
Q&A