Cole-Parmer Narrow-Mouth Plastic Wash Bottle, FEP, 500mL

Representative image only.

Cole-Parmer Narrow-Mouth Plastic Wash Bottle, FEP, 500mL

Cole-Parmer | Item: 0605260

Won't contaminate reagents

Product Description

Key Features

  • Resistant to most chemicals
  • One-piece closure with spout prevents leaks
  • Autoclavable
  • 6 month warranty

More About this Item

FEP bottles resist most chemicals and will not contaminate reagents—use to dispense strong solvents. A one-piece closure with spout prevents leaks.

Certificate of Analysis
For a sample Certificate of Analysis, please go to link below under RESOURCES.
If a Lot specific Certificate of Analysis is required, please call 1-800-323-4340 for assistance.

6 Month Warranty
Specifications
  • Capacity (mL)500
  • Capacity (L)0.5
  • Capacity (oz)16.9
  • Capacity (Gal)0.132
  • MaterialFluorinated Ethylene Propylene (FEP)
  • Cap MaterialFEP
  • Cap / Stopper SizeGL32
  • Cap ColorNatural
  • Bottle ColorNatural
  • Withstands High Temperature (DegreeF)401
  • Withstands Low Temperature (° F)-328
  • AutoclavableYes
  • Neck Inside Diameter (ID) (mm)25
  • Neck Inside Diameter (ID) (in)63/64
  • Outside Diameter (OD) (in)3 5/64
  • Outside Diameter (OD) (cm)7.8
  • Overall Height (in)6 3/8
  • Overall Height (cm)16.2
  • Overall Depth (in)6 19/64
  • Overall Depth (cm)16
  • Overall Width (in)3 5/64
  • Overall Width (cm)7.8
  • DescriptionNarrow-Mouth Plastic Wash bottle, 500ml
  • Warranty6 month
Reviews
Q&A