Cole-Parmer Multiple Rod Connector, Nickel-Plated Zinc, Holds Up to 6 Rod s
Representative image only.
Cole-Parmer Multiple Rod Connector, Nickel-Plated Zinc, Holds Up to 6 Rod s
Cole-Parmer | Item: 0804112
Connect rods and / or clamps to your frame or build a custom lab frame
₹9,700.00INR / Each
₹9,700.00INR / Each
Product Description
Key Features
  • Use with 1/2" (12.7mm) diameter frame rods
  • 1 year warranty
More About this Item
Designed to hold up to six rods allowing for maximum versatility with one connector. The large oval hole in the connector allows for two rods be fitted and adjusting within it. Standard holes accept 1/2" (13mm) rods.
1 Year Warranty
Specifications
  • Clamp MaterialNickel-plated Zinc
  • Clamp TypeFrame and Rod
  • Min Grip Range (InchShort)0
  • Max Grip Range (InchShort)0.5
  • Accepts Rod Diameter (in)0.5
  • DescriptionMultiple Rod Connector, Nickel-Plated Zinc, Holds Up to 6 Rod s