Cole-Parmer Muffle Furnace, 22 L, 1600°C, Silicone Carbide heating element; 1/Ea
Representative image only.
Cole-Parmer Muffle Furnace, 22 L, 1600°C, Silicone Carbide heating element; 1/Ea
Cole-Parmer | Item: 1639403
Continuous use furnaces having vertical lift door that keeps hot surface away when opening
₹5,508,000.00INR / Each
₹5,508,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer Muffle Furnace, 22 L, 1600°C, Silicone Carbide heating element; 1/Ea
Cole-Parmer Muffle Furnace, 22 L, 1600°C, Silicone Carbide heating element; 1/Ea
1639403
₹5,508,000.00INR / Each
+
Cole-Parmer Smoke Extraction Chimney; 1/Ea
1639467
₹68,500.00INR / Each
=
Total Price 5576500.00 ADD 2 ITEMS TO CART
Product Description
Includes
Ceramic tray
Key Features
  • Low thermal mass insulation for low external housing temperature
  • Built with low density ceramic bricks and ceramic fiber for faster heating rate
  • Double insulation includes air chamber lets work at high temperature continuously with air circulation
  • Programmable controller with multiple set points for ramp and dwell options
  • 3 year warranty
More About this Item
Vertical lift door keeps hot surface away when opening. Design is compact and lightweight having operating temperature of 1600 ºC with rapid heating through Kanthal® silicone carbide (SiC) heating elements. Modern design metal case with chrome-phosphatizing base protection and external finish with heat-resistant metal paint. Heat resistance in refractory insulation of very low thermal conductivity coefficient. Door system adjusted on the furnace frame by pressure, allowing for complete sealing. Electrically and thermally insulated door handle. Rear ventilation via ceramic chimney.
These furnaces have lower front control panel, solid state relay, general safety switch and general safety contactor. Each model features PAD Digital control with microprocessor-based temperature controls, PID parameters, non-volatile memory and alarm. Different control options with multiple programming and has data logger and programmer communication by Ethernet/RS-232. These furnaces feature safety shut-off for thermocouple failure and door opening. Different range of accessories available as per the application need.
3 Year Warranty
Specifications
  • DescriptionMuffle Furnace, 22 L, 1600 C, Silicone Carbide heating element; 1/Ea
  • Warranty3 Years