Cole-Parmer Mixer Testing Beads, Yellow Acetate, 3.0 mm Diameter; 2400 Beads/Jar
Representative image only.
Cole-Parmer Mixer Testing Beads, Yellow Acetate, 3.0 mm Diameter; 2400 Beads/Jar
Cole-Parmer | Item: 0456071
Highly visible beads let you easily visualize mixing patterns
₹70,500.00INR / Each
₹70,500.00INR / Each
Product Description
Key Features
  • Calibrated to simulate particles in fluid
  • 1 year warranty
More About this Item
Test your mixer/impeller system for clumping and settling. These highly visible beads are calibrated to simulate particles in your fluid and let you easily visualize mixing patterns. Use different sizes (color-coded) simultaneously to determine effect on different sized particles.
1 Year Warranty
Specifications
  • DescriptionMixer Testing Beads, Yellow Acetate, 3.0 mm Diameter; 2400 Beads/Jar