Cole-Parmer Microscope Slide Mailer, Polypropylene, Holds 1 Slide; 25/Pk
Representative image only.
Cole-Parmer Microscope Slide Mailer, Polypropylene, Holds 1 Slide; 25/Pk
Cole-Parmer | Item: 7890856
Safely transport and protect microscope slides
₹2,400.00INR / Pkg of 25
₹2,400.00INR / Pkg of 25
Product Description
Key Features
  • Constructed of durable polypropylene
  • Holds 3” x 1” (75 x 25 mm) slides
  • Slide mailer holds one 3" x 1" microscope slide
More About this Item
Protect your 75 x 25 mm microscope slides with these durable slide mailers. These white polypropylene mailers feature hinged covers and molded-in dividers to securely hold microscope slides and protect them from damage during transport.
Specifications
  • Slide TypeSlide Mailer
  • SizeHolds 1 slide
  • DescriptionMicroscope Slide Mailer, Polypropylene, Holds 1 Slide; 25/Pk