Cole-Parmer Micro Cup Horn for Ultrasonic Processor
Representative image only.
Cole-Parmer Micro Cup Horn for Ultrasonic Processor
Cole-Parmer | Item: 0471217
₹147,000.00INR / Each
₹147,000.00INR / Each
Product Description
Key Features
  • 3 year warranty
More About this Item
Horn lets you process samples in absolute isolation without probe intrusion. Ideal for pathogenic, radioactive, or biohazardous materials.
Screw cup horn onto your inverted converter then fill with water and suspend test tube inside. Vibrations are transmitted through the water and induce cavitation. ID is 25 mm.
3 Year Warranty
Specifications
  • DescriptionMicro Cup Horn for Ultrasonic Processor