Cole-Parmer Metal Labeling Tape Dispenser, Large; Each
Representative image only.
Cole-Parmer Metal Labeling Tape Dispenser, Large; Each
Cole-Parmer | Item: 0439693
Easy access to high-volume tape from one handy location
₹14,300.00INR / Each
₹14,300.00INR / Each
Product Description
Key Features
  • Metal construction for long-term, durable use
  • Choose from two sizes: small is designed for tape with 1" (2.5 cm) cores, large is compatible with 3" (7.6 cm) cores
  • Each dispenser includes four roll holders for easy refilling
  • Rubber grips on bottom keep dispensers from moving
Specifications
  • MaterialMetal
  • DescriptionMetal Labeling Tape Dispenser, Large; Each