Cole-Parmer MAS250R 250 Ohm Resistor for Current Inputs, 0.1% 0.25 Watt, 20mA Max.
Representative image only.
Cole-Parmer MAS250R 250 Ohm Resistor for Current Inputs, 0.1% 0.25 Watt, 20mA Max.
Cole-Parmer | Item: 8081612
Accept current inputs by connecting an external shunt
₹3,600.00INR / Each
₹3,600.00INR / Each
Product Description
Key Features
  • 250 &Omega precision resistor
  • One is required for each 4 to 20mA input signal on two-channel paperless recorders 80816-00, -02, and -04
  • 1 year warranty
1 Year Warranty
Specifications
  • Compatible WithTwo-channel paperless recorders 80816-00, -02, and -04
  • Description250 Ohm Resistor for Current Inputs, 0.1% 0.25 Watt, 20mA Max.