Cole-Parmer Luer to Plug Fitting, Straight Adapter, White Nylon, Cleanroom Packed, Male Luer Lock; 10/Pk
Representative image only.
Cole-Parmer Luer to Plug Fitting, Straight Adapter, White Nylon, Cleanroom Packed, Male Luer Lock; 10/Pk
Cole-Parmer | Item: 5011050
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
₹1,600.00INR / Pkg of 10
₹1,600.00INR / Pkg of 10
Product Description
Key Features
  • Seal luer ports with a secure and leak-proof connection
  • Luer fittings allow for a simple yet secure twist-lock connection
  • Ideal for capping off fluid transfer system
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Meets FDA regulations 21 CFR 178.3297 and is BSE/TSE and RoHS compliant
More About this Item
These Cole-Parmer® cleanroom fittings are high-quality PVC-free fittings, ideal in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Luer fitting is a standardized system comprised of a male taper and a female mating part that provides a leak-free connection. Simply connect the plug to a female luer adapter to stop the fluid flow. Plugs are available in five different formulations—select one that best suits your application.
Medical nylon fittings are made from the same resin as natural nylon but with FDA-compliant white coloring. They offer good tensile strength, stiffness, and impact resistance and are resistant to fuels and lubricants. Not recommended for use with phenols, strong acids, and oxidizing agents.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialNylon
  • Number Of Ports1
  • Connection TypeMale Luer Lock x Plug
  • Flow PatternStraight
  • Regulatory ApprovalsFDA;Non-Phthalate;RoHS;Non-DEHP
  • Luer Connection Type (Port 1)Male Luer Lock
  • Luer Connection Type (Port 2)Plug
  • DescriptionLuer to Plug Fitting, Straight Adapter, White Nylon, Cleanroom Packed, Male Luer Lock; 10/Pk