Cole-Parmer Luer to Plug Fitting, Straight Adapter, Natural Nylon, Cleanroom Packed, Large Bore Male Luer; 10/Pk
Representative image only.
Cole-Parmer Luer to Plug Fitting, Straight Adapter, Natural Nylon, Cleanroom Packed, Large Bore Male Luer; 10/Pk
Cole-Parmer | Item: 5010949
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
₹3,400.00INR / Pkg of 10
₹3,400.00INR / Pkg of 10
Product Description
Key Features
  • Not compatible with standard luer fittings
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Large-bore luer fittings offer higher flow rates compared to standard luers
  • Natural nylon
  • Complies with FDA 21 CFR 177.1500 (b)(1)
  • RoHS compliant
More About this Item
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Luer fitting is a standardized system comprised of a male taper and a female mating part that provides a leak-free connection. Simply connect the plug to a large-bore female luer adapter to stop the fluid flow. Plugs are available in five different formulations—select one that best suits your application.
Natural nylon fittings provide good tensile strength, stiffness, and impact resistance and are resistant to fuels and lubricants. Not recommended for use with phenols, strong acids, and oxidizing agents.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialNylon
  • Number Of Ports1
  • Regulatory ApprovalsFDA;Non-DEHP;Non-Phthalate;RoHS
  • Luer Connection Type (Port 1)Large Bore Male Luer
  • Luer Connection Type (Port 2)Plug
  • DescriptionLuer to Plug Fitting, Straight Adapter, Natural Nylon, Cleanroom Packed, Large Bore Male Luer; 10/Pk