Cole-Parmer Luer to Plug Fitting, Straight Adapter, Natural Kynar®, Cleanroom Packed, Male Luer Lock; 10/Pk
Representative image only.
Cole-Parmer Luer to Plug Fitting, Straight Adapter, Natural Kynar®, Cleanroom Packed, Male Luer Lock; 10/Pk
Cole-Parmer | Item: 5011051
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
₹1,800.00INR / Pkg of 10
₹1,800.00INR / Pkg of 10
Product Description
Key Features
  • Seal luer ports with a secure and leak-proof connection
  • Luer fittings allow for a simple yet secure twist-lock connection
  • Ideal for capping off fluid transfer system
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Natural Kynar®
  • Biopharma compatible and USP 661 and BPOG compliant
More About this Item
These Cole-Parmer® cleanroom fittings are high-quality PVC-free fittings, ideal in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Luer fitting is a standardized system comprised of a male taper and a female mating part that provides a leak-free connection. Simply connect the plug to a female luer adapter to stop the fluid flow. Plugs are available in five different formulations—select one that best suits your application.
Natural Kynar® fittings provide excellent chemical, flame, gamma radiation, UV, and weather resistance. These robust fittings exhibit good mechanical properties, low moisture absorption, and good machinability. Natural Kynar®, also known as natural Polyvinylidene Fluoride (PVDF), is a PFAS-free resin.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialPVDF
  • Number Of Ports1
  • Connection TypeMale Luer Lock x Plug
  • Flow PatternStraight
  • Regulatory ApprovalsNon-Phthalate;USP;Non-DEHP
  • Luer Connection Type (Port 1)Male Luer Lock
  • Luer Connection Type (Port 2)Plug
  • DescriptionLuer to Plug Fitting, Straight Adapter, Natural Kynar®, Cleanroom Packed, Male Luer Lock; 10/Pk