Cole-Parmer Luer to Plug Fitting, Straight Adapter, CrystalVu™, Cleanroom Packed, Large Bore Male Luer; 10/Pk
Representative image only.
Cole-Parmer Luer to Plug Fitting, Straight Adapter, CrystalVu™, Cleanroom Packed, Large Bore Male Luer; 10/Pk
Cole-Parmer | Item: 5010946
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
₹3,900.00INR / Pkg of 10
₹3,900.00INR / Pkg of 10
Product Description
Key Features
  • Not compatible with standard luer fittings
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Large-bore luer fittings offer higher flow rates compared to standard luers
  • CrystalVu™
  • USP Class VI, EU 2002/72/EC, FDA 21 CFR section 177.1520, and RoHS compliant
More About this Item
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Luer fitting is a standardized system comprised of a male taper and a female mating part that provides a leak-free connection. Simply connect the plug to a large-bore female luer adapter to stop the fluid flow. Plugs are available in five different formulations—select one that best suits your application.
A transparent alternative to polycarbonate, CrystalVu™ fittings do not contain any bisphenol-A (BPA) that has been found to be harmful to humans. These fittings have excellent chemical resistance, withstand temperatures up to 180°C (356°F), and can be autoclaved multiple times with no crazing.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialPolycarbonate
  • Number Of Ports1
  • Regulatory ApprovalsNon-DEHP;Non-Phthalate;FDA;RoHS;USP
  • Luer Connection Type (Port 1)Large Bore Male Luer
  • Luer Connection Type (Port 2)Plug
  • DescriptionLuer to Plug Fitting, Straight Adapter, CrystalVu™, Cleanroom Packed, Large Bore Male Luer; 10/Pk