Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
Free technical application support available to determine which connector type is needed for your application
PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
Forms a leak-free luer connection with a quick simple turn
Single barb design tightly “grips” the tubing
White nylon
RoHS compliant
Female luer x 1/8" hose barb
More About this Item
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Quickly and easily transition from soft tubing to a luer connection using these adapters. Luer fitting is a standardized system comprised of a male taper and a female mating part that provides a leak-free connection. A single barb on one end of the fitting creates a tight seal to keep the tubing in place.
White nylon fittings provide good tensile strength, stiffness, and impact resistance and are resistant to fuels and lubricants. They contain recycled runner material produced during virgin material, natural nylon production runs. Not recommended for use with phenols, strong acids, and oxidizing agents.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
Fitting MaterialNylon
Connection TypePanel-Mount Female Luer x Hose Barb
Number Of Ports2
Barb Size / Tubing Id (Port 2) (in)1/8
Regulatory ApprovalsNon-Phthalate;RoHS;Non-DEHP
Luer Connection Type (Port 1)Female Luer
DescriptionLuer to Hose Barb Fitting, Straight Adapter, White Nylon, Cleanroom Packed, Panel-Mount Female Luer x 1/8" ID; 10/Pk