Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
Free technical application support available to determine which connector type is needed for your application
PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
Forms a leak-free luer connection with a quick simple turn
Single barb design tightly “grips” the tubing
Biopharma compatible and USP 661 and BPOG compliant
Female luer x 1/16" hose barb
More About this Item
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Quickly and easily transition from soft tubing to a luer connection using these adapters. Luer fitting is a standardized system comprised of a male taper and a female mating part that provides a leak-free connection. A single barb on one end of the fitting creates a tight seal to keep the tubing in place. These natural Kynar® fittings provide excellent chemical, flame, gamma radiation, UV, and weather resistance. They also exhibit good mechanical properties, low moisture absorption, and good machinability. Natural Kynar®, also known as natural Polyvinylidene Fluoride (PVDF), is a PFAS-free resin.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
Fitting MaterialPVDF
Connection TypeFemale Luer x Hose Barb
Number Of Ports2
Barb Size / Tubing Id (Port 2) (in)1/16
Flow PatternStraight
Regulatory ApprovalsNon-DEHP;USP;Non-Phthalate
Luer Connection Type (Port 1)Female Luer
Luer Connection Type (Port 2)Hose Barb
DescriptionLuer to Hose Barb Fitting, Straight Adapter, Natural Kynar®, Cleanroom Packed, Female Luer x 1/16" ID; 10/Pk